missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ Amber Glass E-Z Vials

Manufacturer:  DWK Life Sciences W2251730204

 View more versions of this product

Catalog No. 02-926-954


 

Description

Description

  • Manufactured from certified clear and amber Type I borosilicate glass
  • Vials are packaged 100 per shelf pack and 10 packs per case
  • 40% larger opening improves sample accessibility while reducing autosampler needle damage
Specifications

Specifications

E-Z Vial™ Pack
1.8mL
Blue
32mm
12mm
Yes
Aluminum
11mm
Borosilicate Glass
Crimp
100 Pack
1.25 in.
PTFE/Silicone
1.8mL
Documents