Please login to your online account to display your discounted pricing
Wheaton™ Amber Boston Rounds without Caps
Amber glass protects light-sensitive solutions
$176.37 - $507.40
Specifications
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Screw
Closure Color White
Color Amber
View More Specs
Products
Catalog Number Mfr. No. Capacity Diameter (English) Height (English) Closure Size Price Quantity    
02-912-165
wheaton™
W216866
2 oz., 60mL 1.53 in. 3.7 in. 20-400 Case of 288 Each for $434.80
02-912-164
wheaton™
W216865
1 oz., 30mL 1.22 in. 3.1 in. 20-400 Case of 432 Each for $507.40
02-912-169
wheaton™
W216870
32 oz., 1000mL 3.7 in. 8.1 in. 33-400 Case of 30 Each for $176.37
02-912-167
wheaton™
W216868
8 oz., 250mL 2.36 in. 5.3 in. 24-400 Case of 108 Each for $251.51
02-912-166
wheaton™
W216867
4 oz., 125mL 1.88 in. 4.4 in. 22-400 Case of 160 Each for $317.40
02-912-168
wheaton™
W216869
16 oz., 500mL 2.95 in. 6.6 in. 28-400 Case of 60 Each for $210.60
Description & Specifications
Specifications
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Screw
Closure Color White
Color Amber
Material Soda Lime Glass
  • Type III soda-lime glass
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.