Please login to your online account to display your discounted pricing
Wheaton™ Amber Boston Rounds with Rubber-lined Caps
Amber glass protects light-sensitive solutions
$51.31 - $105.34
Specifications
Closure Liner Rubber
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Color Amber
View More Specs
Products
Catalog Number Mfr. No. Capacity Closure Size Diameter (English) Height (English) Price Quantity    
02-911-368
wheaton™
W216854
2 oz., 60mL 20-400 1.22 in. 3.1 in. Case of 24 Each for $70.37
02-911-369
wheaton™
W216855
4 oz., 125mL 22-400 1.88 in. 4.4 in. Case of 24 Each for $77.20
02-911-370
wheaton™
W216856
8 oz., 250mL 24-400 2.36 in. 5.43 in. Case of 12 Each for $51.31
02-911-371
wheaton™
W216857
16 oz., 500mL 28-400 2.95 in. 6.6 in. Case of 12 Each for $66.23
02-911-372
wheaton™
W216858
32 oz., 1000mL 33-400 3.7 in. 8.18 in. Case of 12 Each for $105.34
Description & Specifications
Specifications
Closure Liner Rubber
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Color Amber
Material Glass
  • Type III soda-lime glass