missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ Amber Boston Rounds with Polyvinyl-lined Caps

Amber Type III soda lime glass protects light-sensitive solutions

Manufacturer:  DWK Life Sciences W216836

 View more versions of this product

Catalog No. 02-911-350


 

Description

Description

  • Available in partitioned Convenience Pack or Valu-Bulk™ cartons
  • Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage

Each case of Convenience Pack bottles includes Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage

Order Info

Caps can be ordered separately.

Compliance

USP Type III soda-lime glass

Specifications

Specifications

1 oz., 30mL
1 oz.
Poly/Vinyl
1.22 in.
Soda Lime Glass
White
31mm
Screw
30mL
Polypropylene
Amber
3.1 in.
Convenience pack
20-400
79mm
Documents