missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Amber Boston Rounds with Polyvinyl-lined Caps
Amber Type III soda lime glass protects light-sensitive solutions
Supplier: DWK Life Sciences W216863
Description
Amber Type III soda lime glass protects light-sensitive solutions
- Available in partitioned Convenience Pack or Valu-Bulk™ cartons
- Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage
Each case of Convenience Pack bottles includes Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage
Order Info
Caps can be ordered separately.
Compliance
USP Type III soda-lime glass
Specifications
Polypropylene | |
75 mm | |
Screw Cap | |
28-400 mm | |
500 mL | |
Amber | |
Soda Lime Glass | |
White |
168 mm | |
6.6 in. | |
Poly/Vinyl | |
16 oz., 500 mL | |
16 oz. | |
2.95 in. | |
Valu-bulk pack |