Please login to your online account to display your discounted pricing
Wheaton™ Amber Boston Rounds with PE Cone Lined Caps
Amber Type III soda lime glass protects light-sensitive solutions
$65.11 - $107.63
Specifications
Closure Liner Polyethylene
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Color Amber
View More Specs
Products
Catalog Number Mfr. No. Capacity Closure Size Diameter (English) Height (English) Price Quantity    
02-911-362
wheaton™
W216848
1 oz., 30mL 20-400 1.22 in. 3.1 in. Case of 48 Each for $107.63
02-911-363
wheaton™
W216849
2 oz., 60mL 20-400 1.53 in. 3.7 in. Case of 24 Each for $66.91
02-911-364
wheaton™
W216850
4 oz., 125mL 22-400 1.88 in. 4.4 in. Case of 24 Each for $81.74
02-911-365
wheaton™
W216851
8 oz., 250mL 24-400 2.36 in. 5.43 in. Case of 12 Each for $65.11
02-911-366
wheaton™
W216852
16 oz., 500mL 28-400 2.95 in. 6.6 in. Case of 12 Each for $65.54
02-911-367
wheaton™
W216853
32 oz., 1000mL 33-400 3.7 in. 8.18 in. Case of 12 Each for $76.60
Description & Specifications
Specifications
Closure Liner Polyethylene
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Color Amber
Material Soda Lime Glass
  • Narrow mouth enables liquids to be carefully poured to prevent spillage
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.