missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wheaton™ Amber Boston Rounds with PE Cone Lined Caps

Amber Type III soda lime glass protects light-sensitive solutions

Manufacturer:  Wheaton™ W216849

 View more versions of this product

Catalog No. 02-911-363


 

Specifications Description & Specifications

Specifications

2 oz., 60mL
2 oz.
Polyethylene
1.53 in.
Soda Lime Glass
20-400
94mm
60mL
Phenolic
Amber
3.7 in.
Black
39mm
Screw