DWK Life Sciences Wheaton™ Perforated Aluminum Capsules
Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.
Manufacturer:  DWK Life Sciences 22419306
 View more versions of this product
Catalog No. 06-451-85
Description
Description
Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.
  • Good barrier properties
  • Color selection includes natural, red, blue and green
  • Chemically resistant to alcohol and acetone, among many others
Specifications
Specifications
Red
Aluminum
1000/Cs.
20 mm
Complete Tear-off
SDS
Documents
Product Certifications
Accessories