DWK Life Sciences Wheaton™ Center Disc Tear-Out Aluminum Seals, Unlined
Tear-out seals remove completely allowing content to pour from bottle. Wheaton™ Center Disc Tear-Our Aluminum Seals, Unlined are available in a wide range of sizes and colors.
Manufacturer:  DWK Life Sciences 22418301
 View more versions of this product
Catalog No. 06-406-14B
Description
Description
  • Good barrier properties
  • Color selection includes natural, red, blue and green
  • Chemically resistant to alcohol and acetone, among others
Specifications
Specifications
Natural
Aluminum
1000/Cs.
20 mm
Center Disc Tear-out