DWK Life Sciences Wheaton™ Center Disc Tear-Out Aluminum Seals, Unlined
Tear-out seals remove completely allowing content to pour from bottle. Wheaton™ Center Disc Tear-Our Aluminum Seals, Unlined are available in a wide range of sizes and colors.
Supplier: DWK Life Sciences 22418701
Description
- Good barrier properties
- Color selection includes natural, red, blue and green
- Chemically resistant to alcohol and acetone, among others
Specifications
Natural | |
1000/Cs. | |
30 mm |
Aluminum | |
Center Disc Tear-out |