Please login to your online account to display your discounted pricing
Wheaton™ Acura™ electro 956 Eight-Channel Pipets
Fits any single channel volumetric module
Manufacturer: wheaton™  WW870930
 View more versions of this product
Catalog No. 03-410-115
  • / Each
Description & Specifications
Specifications
Number of Channels 8.0
Volume (Metric) Range 20 to 350μL
Compatible Tips Universal fit pipette tip
Increments 5μL
Model Acura™ Electro™ 956
Warranty 2 Years
Display Digital
Accuracy ±1 to <±0.6%
Includes Microprocessor controlled motor, rechargeable battery pack, QC certificate, operating instructions and samples of pipette tip
Battery Type Rechargeable
Volume (Metric) 20 to 350μL
Tip Style 350μL
Finger Style Ergonomic