Please login to your online account to display your discounted pricing
Wheaton™ Acura™ Electro 926XS Single Channel Micropipet
Unsurpassed metrological performance and stability
Manufacturer: wheaton™  WW870902
 View more versions of this product
Catalog No. 03-410-101
  • / Each
Description & Specifications
Specifications
Number of Channels 1
Programmability Self Teaching
Warranty 2 Years
Finger Style Ergonomic
Model 926XS
Increments 0.01μL
Battery Type NiMH
Display Digital
Compatible Tips Ultra 10μL pipette tip
Includes Rechargeable battery pack, individual QC certificate, pipette tip samples and operating instructions
Accuracy ±2.5 to <±0.9%
For Use With (Equipment) Ultra 10μL pipette tip
Volume (Metric) Range 0.1 to 2μL
Volume (Metric) 0.1 to 2μL