Please login to your online account to display your discounted pricing
Wheaton™ Acura Electro™ 926 XS and 936 Single Channel Pipette - Basic Package
Includes a microprocessor-controlled motor and rechargeable battery pack
Manufacturer: wheaton™  WW870902A
 View more versions of this product
Catalog No. 03-410-001
  • / Case
Description & Specifications
Specifications
Number of Channels 1
Volume (Metric) 0.1 to 2μL
Compatible Tips Ultra 10μL pipette tip
Warranty 2 Years
Increments 0.01μL
Includes Electronic pipette, individual QC certificate and operating instructions, pipette charging stand, fast charger with AC adapter and cord, microprocessor controlled motor, additional battery pack and pipette tip samples
For Use With (Equipment) Ultra 10μL pipette tip
Model 926XS
Display Digital
Finger Style Ergonomic
Volume (Metric) Range 0.1 to 2μL
Accuracy ±2.5 to <±0.9%
Battery Type NiMH