Please login to your online account to display your discounted pricing
Wheaton™ Acura Electro™ 926 XS and 936 Single Channel Pipette - Basic Package
Includes a microprocessor-controlled motor and rechargeable battery pack
Manufacturer: wheaton™  W870910A
 View more versions of this product
Catalog No. 03-410-005
  • / Case
Description & Specifications
Specifications
Number of Channels 1
Number of Channels 1
Accuracy ±1 to <±0.5%
Battery Type NiMH
Compatible Tips 200μL Pipette tip
Display Digital
Finger Style Ergonomic
For Use With (Equipment) 200μL Pipette tip
Includes Electronic pipette, individual QC certificate and operating instructions, pipette charging stand, fast charger with AC adapter and cord, microprocessor controlled motor, additional battery pack and pipette tip samples
Increments 0.25μL
Warranty 2 Years
Model 926XS
Volume (Metric) Range 2.5 to 50μL
Volume (Metric) 2.5 to 50μL
Capacity Range 2.5 to 50μL
Compatibility 200μL Pipette tip