Wheaton™ AC Medium Rounds: No Caps
Ideal for use as sample bottles
$145.74 - $296.86
Specifications
Closure Type Screw
Color Clear
Material Soda Lime Glass
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Closure Size Diameter (English) Height (English) Price Quantity  
Catalog Number Mfr. No. Capacity Closure Size Diameter (English) Height (English) Price Quantity  
02-912-240 Wheaton™
W217005
1 oz., 30mL 33-400 1.33 in. 2.67 in. Case of 432 Each for $296.86
02-912-241 Wheaton™
W217006
2 oz., 60mL 38-400 1.65 in. 3.26 in. Case of 288 Each for $254.20
02-912-243 Wheaton™
W217008
8 oz., 250mL 58-400 2.44 in. 5 in. Case of 96 Each for $147.31
02-912-242 Wheaton™
W217007
4 oz., 125mL 48-400 2 in. 4.01 in. Case of 144 Each for $208.03
02-912-244 Wheaton™
W217009
16 oz., 500mL 70-400 2.99 in. 5.7 in. Case of 48 Each for $145.74
Specifications
Description
  • Manufactured from Wheaton 800 glass, which meets USP Type III standards
  • Useful for dry powders, soil samples, and specimen storage
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Screw
Clear
Soda Lime Glass