missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wheaton™ ABC Vial Convenience Packs: Clear Glass

40% larger opening improves sample accessibility while reducing autosampler needle damage

Manufacturer:  Wheaton™ W22515002

 View more versions of this product

Catalog No. 03-410-208


 

Specifications Description & Specifications

Specifications

1.8mL
12mm
32mm
PTFE/Silicone
9mm
Screw cap
Natural
0.47 in.
1.25 in.
Polypropylene
Chromatography Vial
1.8mL
Borosilicate Glass