missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wheaton™ ABC Vial Convenience Packs: Clear Glass

40% larger opening improves sample accessibility while reducing autosampler needle damage

Manufacturer:  Wheaton™ WW22515501

 View more versions of this product

Catalog No. 02-912-913


 

Specifications Description & Specifications

Specifications

1.8mL
12mm
32mm
Polypropylene
Chromatography Vial
1.8mL
Screw cap
Natural
0.47 in.
1.25 in.
Yes
PTFE/Red Silicone
9mm
Natural
Borosilicate Glass