Please login to your online account to display your discounted pricing

Wheaton™ ABC Vial Caps


Polypropylene screw caps for use with ABC 12 x 32mm vials

Manufacturer: wheaton™  WW22533001

 View more versions of this product

Catalog No. 02-912-901

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Type Screw Cap
Material (Liner) PTFE/Silicone
Color Natural
Material Closure Polypropylene
Thread Size 9mm
Material Polypropylene