Please login to your online account to display your discounted pricing

Wheaton™ ABC Vial Caps


Polypropylene screw caps for use with ABC 12 x 32mm vials

Manufacturer: wheaton™  WW2253300101

 View more versions of this product

Catalog No. 02-912-902

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Material Polypropylene
Type Screw Cap
Color Black
Material (Liner) PTFE/Silicone
Material Closure Polypropylene
Thread Size 9mm