DWK Life Sciences Wheaton™ 2D Data Matrix Bar Code Bottom Insert
For use with Wheaton CryoELITE™ Freestanding Cryogenic Vials
Supplier:  DWK Life Sciences W2246210303
 View more versions of this product
Catalog No. 03-410-320
Description
Description
  • Makes sample identification and traceability reliable and convenient
  • Bar code insert manually press fits into bottom of vial
  • Data matrix readable with standard scanning hardware
  • Made from same cryogenic radiation-grade, virgin polypropylene as vial, ensuring uniform thermal expansion
Specifications
Specifications
Polypropylene
Sterile
500/Cs.