DWK Life Sciences Wheaton™ 25mm Filtration Assembly With No. 5 Stopper Connection With Stainless Steel Support
Designed to handle small volumes of liquids for analysis of particulate or microbiological contamination
Supplier:  DWK Life Sciences 419327
 View more versions of this product
Catalog No. 14209105
Includes:
  • 316 stainless steel support with 120 mesh screen
  • PTFE support screen gasket
  • Glass support base
  • 15mL graduated funnel
  • Anodized aluminum clamp
  • No. 5 silicone stopper
Description
Description
  • For filtration of viscous or proteinaceous solutions to produce ultra clean filtrate
  • Glass components manufactured from low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Specifications
Specifications
USP Type I, ASTM E 438 Type I, Class A
Borosilicate Glass
One 316 stainless steel support with 120 mesh screen, PTFE support screen gasket, glass support base, 15mL graduated funnel, anodized aluminum clamp, one No. 5 silicone stopper
Filtration Assembly
25mm filtration assembly
Filtering viscous or proteinaceous solutions, to produce ultraclean filtrate
1