missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wheaton™ 12 x 32mm Glass/Plastic Vials with 0.1mL Limited Volume Insert

Limited volume, glass/plastic vials

Manufacturer:  Wheaton™ 225190

 View more versions of this product

Catalog No. 03-336-7


 

Specifications Description & Specifications

Specifications

Autosampler Vial
0.1mL
Clear
32mm
0.1mL glass insert
1.25 in.
11mm
Glass, Plastic
Snap/Crimp
100 Case
12mm
0.1mL