Whatman™ Benchkote™ Surface Protectors: PROMO OFFER
Suitable for saturation with disinfectant
$70.73 - $70.73
Specifications
Material Polyethylene
Absorbancy 0.75L/m2
For Use With (Application) Suitable for saturation with disinfectant
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Dimensions (L x W) Quantity Product Type Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Quantity Product Type Price Quantity  
12007180PM Whatman™
2300916PM
18 x 22.5 in. (460 x 570mm) 50 Pack Benchtop Protector - Sheets Pack of 50 Each for $70.73
Specifications
Description
  • High-quality absorbent paper coated on one side with an impermeable layer of polyethylene that prevents liquids from reaching benchtops
  • Paper side absorbs solvents, toxic liquids, pathogens and radiochemicals and can be written on with ink or pencil
  • Benchkote Plus™ sheets and rolls have a thicker absorbent layer than standard Benchkote protectors and can absorb more than 0.75L/m2
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
Description & Specifications
Specifications
Polyethylene
0.75L/m2
Suitable for saturation with disinfectant