Whatman™ Vacuum-Type Glass Membrane Holder
Produced from borosilicate glass and available with a choice of support screen. Whatman™ Vacuum-Type Glass Membrane Holder is suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.
Manufacturer:  Whatman™ 1960009
 View more versions of this product
Catalog No. 09-907-3
Specifications
Description
Holder, Filter; Whatman; Vacuum; Reservoir Capacity: 1000mL; Filter Diameter: 90mm; Glass Support Screen
  • Made from borosilicate glass
  • Ensures secure clamping and integral sealing at membrane edge
  • Gravimetric analysis filtration
  • Ultraclean filtration
  • Bacteriological control of food, drink, pharmaceuticals, cosmetics
  • Removal of particulates in solution
  • Nucleic acid and protein analysis
  • HPLC solvent filtration
Description & Specifications
Specifications
Filter Holder
Borosilicate Glass
Prefilter Membrane: 70mm dia.
38.5cm2
Produced from borosilicate glass and available with a choice of support screen. Whatman™ Vacuum-Type Glass Membrane Holder is suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.
90mm
1L