Please login to your online account to display your discounted pricing

Whatman™ UniPrep™ Syringeless Filters - Nylon

Process samples in one-third the time of conventional syringe filters

$426.54 - $5,377.26

Specifications

Diameter (Metric) 17mm
Type Syringeless Filter
Filtration Area (Metric) 0.28cm sq.
Material Nylon (NYL)
View More Specs
Products
Catalog Number Mfr. No. Pore Size Quantity Price Quantity    

09-923-7

 
whatman™
UN113UNYL
0.45μm Pk. of 50 Pack of 50 Each for $426.54

09-923-8

 
whatman™
WHTUN113ENYL
0.2μm Pk. of 50 Pack of 50 Each for $426.54

09-923-9A

 
ge healthcare - whatman
UN513UNYL
0.45μm Pk. of 1000 Pack of 1000 Each for $5,377.26
Description & Specifications

Specifications

Diameter (Metric) 17mm
Type Syringeless Filter
Filtration Area (Metric) 0.28cm sq.
Material Nylon (NYL)

UniPrep syringeless filters are preassembled filtration devices for the filtration and storage of laboratory samples.

  • Naturally hydrophilic membrane for filtration of samples containing aqueous or organic solvents with a pH range between 3 to 10
  • Consists of external sample test tube, plunger (which incorporates membrane filter and prefilter), removable cap
  • Filtration area: 0.3cm2
  • Safe, efficient—prefilter and membrane can't pop off
  • Air purge unnecessary—inserting plunger into tube provides a positive pressure purge of sample through filter
  • Sealed glass microfiber prefilter traps larger particles before they can clog the membrane

  • Increase lab productivity and efficiency
  • Generate less waste
  • Realize greater overall cost savings per test