Whatman™ UNIPLATE™ 96-Well Clear Microplates
One-piece molded collection plates designed to suit a variety of applications
$353.17 - $353.17
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
PF77011651 Whatman™
77011651
Pack of 50 Each for $353.17
Specifications
Description
  • Polystyrene
  • Withstand low temperatures (-20°C) for long-term storage
  • Flat, Round or V-shaped well bottoms
  • Conform to ANSI™/SBS (American National Standards Institute, Society of Biomolecular Screening) standards for compatibility with most microplate readers and automated plate-handling devices
  • Untreated
Black and white UNIPLATE microplates are also available.