Please login to your online account to display your discounted pricing
Whatman™ UniFilter™ Microplates, 96-Well, 2mL
Use for biomolecular purification by solid-phase extraction and organic synthesis in combinatorial chemistry library generation
Manufacturer: whatman™  77007201
 View more versions of this product
Catalog No. PF77007201
  • / Pack of 25 Each
Description & Specifications
Specifications
For Use With (Application) Use for biomolecular purification by solid-phase extraction and organic synthesis in combinatorial chemistry library generation, sample preparation, genomics, filter-based assays and SPE
Format 96-well
Includes Long drip director
Material Glass-Filled Polypropylene
Membrane GF/C
No. of Wells 96
Type Filtering Microplate
Volume (Metric) Well 2000μL
Volume (Metric) Well 2000μL