Whatman™ UniFilter™ Microplates, 96-Well, 2mL
Use for biomolecular purification by solid-phase extraction and organic synthesis in combinatorial chemistry library generation
$1,168.11 - $1,168.11
Specifications
Format 96-well
Material Glass-Filled Polypropylene,Glass-Filled Polypropylene
Type Filtering Microplate
For Use With (Application) Use for biomolecular purification by solid-phase extraction and organic synthesis in combinatorial chemistry library generation, sample preparation, genomics, filter-based assays and SPE
No. of Wells 96,96
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Membrane Price Quantity  
Catalog Number Mfr. No. Membrane Price Quantity  
PF77007201 Whatman™
77007201
GF/C Pack of 25 Each for $1,168.11
Specifications
Description
  • Glass-filled polypropylene microplates
  • Patented integral filter design prevents well-to-well cross contamination
  • Standard footprint—compatible with most plate readers, autosamplers and robots
  • Unique drip director design on the bottom of the plates controls filtrate placement
  • Available in several media types
Description & Specifications
Specifications
96-well
Filtering Microplate
96,96
Glass-Filled Polypropylene,Glass-Filled Polypropylene
Use for biomolecular purification by solid-phase extraction and organic synthesis in combinatorial chemistry library generation, sample preparation, genomics, filter-based assays and SPE
2000μL