Cytiva Whatman™ UniFilter™ 384-Well Microplates
Microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples.
Supplier: Cytiva 77001101
Description
Microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples.
Specifications
Filter | |
Whatman GF/C | |
16 x 24 | |
Polystyrene | |
Can be used with gel filtration media for high-throughput sequencing reaction clean-up, including removal of dye blobs | |
50/Pk. |
Standard | |
GEâs Whatman UNIFILTER 384 well microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples. | |
Clear | |
384 | |
Long drip director |