Please login to your online account to display your discounted pricing

Whatman™ UniFilter™ 384-Well Microplates

Long drip directors eliminate well-to-well contamination during filtration

$2,642.09 - $2,844.74

Specifications

Color Clear
Format 384-well
Material Polystyrene
Type Filtering Microplate
For Use With (Application) Ideal for use in DNA template cleanup, cell capture and the removal of unwanted debris
View More Specs
Products
Catalog Number Mfr. No. Membrane Price Quantity    

PF77001101

 
whatman™
77001101
Whatman GF/C Pack of 50 Each for $2,642.09

36-103-0948

 
whatman™
77002106
0.45 Hydrophilic PVDF Pack of 50 Each for $2,844.74
Description & Specifications

Specifications

Color Clear
Format 384-well
Material Polystyrene
Type Filtering Microplate
For Use With (Application) Ideal for use in DNA template cleanup, cell capture and the removal of unwanted debris
Includes Long drip director
No. of Wells 384
Volume (Metric) Well 100μL

  • Clear polystyrene construction
  • Ideal for use in DNA template cleanup, cell capture, and the removal of unwanted debris
  • 384-well format microplates with a 100μL well volume allow a large enough sample for recovery after filtration

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.