Please login to your online account to display your discounted pricing
Uni-Core Punch
Disposable punch for use with FTA cards, FTA Elute Cards, and EasiCollect device
Manufacturer: ge healthcare - whatman  WB100082
 View all options for this product
Catalog No. 09-301-278
Description & Specifications
Diameter (Metric) 6mm
For Use With (Application) Whatman™ FTA™ cards, FTA™ elute cards and EasiCollect device
Includes Two cutting mats
Product Type Accessory, FTA Card
Quantity 25 Pack
  • Punch with polished steel tip that is case-hardened and can be sterilized
  • Stainless steel cutting edges enable manual coring, retrieval, and controlled ejection of FTA punch
  • Housed in plastic handle
  • Each tip provides up to 500 punches before disposal
  • Available in multiple diameters varying from 0.35 to 12mm
  • Manual punching with low sample volumes punching 1 to 50 samples per day
  • Minimum contamination and carryover between samples when cleaning procedure is followed