missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GE Healthcare Whatman™ Uni-Core Punch

Disposable punch for use with FTA cards, FTA Elute Cards, and EasiCollect device

Manufacturer:  GE Healthcare WB100039

 View more versions of this product

Catalog No. 05-715-109



Description

Description

  • Punch with polished steel tip that is case-hardened and can be sterilized
  • Stainless steel cutting edges enable manual coring, retrieval, and controlled ejection of FTA punch
  • Housed in plastic handle
  • Each tip provides up to 500 punches before disposal
  • Available in multiple diameters varying from 0.35 to 12mm
  • Manual punching with low sample volumes punching 1 to 50 samples per day
  • Minimum contamination and carryover between samples when cleaning procedure is followed

Specifications

Specifications

0.11 in.
Whatman™ FTA™ cards, FTA™ elute cards and EasiCollect device
Accessory, FTA Card
3.0mm
Two cutting mats
4 Pack
Documents