Please login to your online account to display your discounted pricing

Whatman™ Type WCN Cellulose Nitrate Membranes


Recommended for majority of routine applications, this membrane is manufactured under strictly controlled conditions.  GE Healthcare Type WCN Cellulose Nitrate Membranes narrow pore size distribution for improved surface capture and analysis.

Manufacturer: whatman™  WHT7184001

 View more versions of this product

Catalog No. 09-905-14

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Pore Size 0.45μm
Material Cellulose Nitrate
Diameter (Metric) Outer 13mm
Color White
Autoclavable Yes
Quantity (No. per Case) 100/pk.