Cytiva Whatman™ TurboBlotter Transfer System, Small Refills
For use with TurboBlotter Kits
Supplier:  Cytiva 10416306
Catalog No. 36-100-5810
Description
Description
Refill packs contain additional membranes and papers needed for further transfers.
  • Consists of one sheet of Nytra™ SPC 0.45μm Nylon membrane
  • One wick of 3MM Chr blotting paper
  • Blotter stack composed of eight sheets of 3MM Chr blotting paper
  • 20 sheets of GB004 blotting paper
  • Refills contain five sets of transfer materials