Whatman™ TurboBlotter Transfer System, Large Kits
Downward blotting device turns the conventional Northern/Southern transfer stack upside down for rapid, high-resolution transfer of DNA and RNA
$547.48 - $707.21
Specifications
Includes 21 x 26cm Large transfer device, 5 sets of transfer materials (set consists of one sheet of Nytrantrademark (TM) SPC 0.45Glc mum Nylon membrane, one wick of 3mm chromatography blotting Paper and a blotter stack composed of eight sheets o
View More Specs
Includes: Includes device plus five sets of transfer materials for gel size (21 x 26cm) indicated in ordering table. Each set consists of one sheet of Nytrantrademark (TM) SPC 0.45Glc mum Nylon membrane, one wick of 3MM Chr blotting paper and a blotter stack composed of eight sheets of 3MM Chr blotting paper and 20 sheets of GB004 blotting paper. Refills, available separately, contain five sets of transfer materials.
Products ${productFamilyLength}
Catalog Number Mfr. No. Length (English) Gel Length (Metric) Gel Width (English) Gel Width (Metric) Gel Price Quantity  
Catalog Number Mfr. No. Length (English) Gel Length (Metric) Gel Width (English) Gel Width (Metric) Gel Price Quantity  
09-301-184 Whatman™
WHT10416316C
7.8 in. 20cm 5.9 in. 15cm Each for $547.48
09-301-188 Whatman™
WHT10416324C
9.8 in. 25cm 7.8 in. 20cm Each for $707.21
Specifications
Description
  • Takes advantage of gravity — no heavy weights are required on the top of the transfer stack, eliminating the messy set-up of standard upward capillary transfers
  • Has a smaller footprint than most homemade devices and is stackable — up to five units can be stacked during transfers
  • Allows for alkaline buffer transfers in one hour and neutral (SSC) transfers in three hours
  • Very easy to set up and works without power or vacuum source
  • Requires less buffer and blotting paper than conventional transfers
  • Reusable
  • Model is 21 × 26cm, for gels from 12 × 21cm to 20 × 25cm
Order Info
Refill packs containing additional membranes and papers required for further transfer are available separately (Cat. Nos. 09-301-177, -179, -181, -183, -185 and -189).
Description & Specifications
Specifications
21 x 26cm Large transfer device, 5 sets of transfer materials (set consists of one sheet of Nytrantrademark (TM) SPC 0.45Glc mum Nylon membrane, one wick of 3mm chromatography blotting Paper and a blotter stack composed of eight sheets o