Whatman™ Shark Skin Filter Paper
High wet strength allows use under pressure
$66.35 - $132.36
Specifications
Format Circle
Material Cellulose
Flow Rate Medium Slow
Thickness 170μm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
36-100-5713 Whatman™
WHT10347523
385mm Pack of 100 Each for $132.36
09-924-398 Whatman™
10347530
320mm Pack of 100 Each for $116.62
09-924-200 Whatman™
10347577
290mm Pack of 100 Each for $100.77
09-924-388 Whatman™
10347509
90mm Pack of 100 Each for $66.35
09-924-392 Accessories Whatman™
10347513
150mm Pack of 100 Each for $85.09
09-924-390 Accessories Whatman™
10347511
125mm Pack of 100 Each for $103.47
09-924-389 Whatman™
10347510
110mm Pack of 100 Each for $68.85
09-924-394 Whatman™
WHT10347521
270mm Pack of 100 Each for $114.00
09-924-393 Whatman™
10347519
240mm Pack of 100 Each for $95.16
09-924-395 Whatman™
10347522
340mm Pack of 100 Each for $119.38
09-924-397 Whatman™
10347525
500mm Pack of 100 Each for $125.96
Specifications
Description
  • With crepe surface
  • Material: cellulose
  • White circles
  • Flow rate: medium to slow
  • Particle retention: 8-12μm
  • Typical thickness: 170μm
  • Filtration speed: 77.5s
  • Resistant to weak acids and bases
  • Used as protective paper for filter press cloths and in processing of cocoa butter and edible oils
Description & Specifications
Specifications
Circle
Cellulose
Medium Slow
170μm