Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ Sealable Hybridization Bags
Make hybridizations and incubations simpler
Manufacturer:  Cytiva (Formerly GE Healthcare Life Sciences) 10483064
Catalog No. 09927752
Description
Description
Make hybridizations and incubations simpler
Use for hybridization and incubation reactions on thin wet membranes.
  • Open on three sides for easy insertion of wet membrane
  • Can be sealed with a standard heat-sealer
  • Size: 20.3 × 25.8cm (8 × 10 in.)
Specifications
Specifications
50 Pack
Hybridization and incubation reaction