Please login to your online account to display your discounted pricing
Whatman™ 25mm Roby™ Syringe Filters
Developed specifically for automated sample filtration systems. Whatman™ Roby™ 25mm Syringe Filters are optimized for main tablet testing instruments.
$1,031.00 - $3,993.28
Specifications
Diameter (Metric) 25mm
Type Syringe Filter
Inlet Style Female luer lock
Outlet Style Male luer
View More Specs
Products
Catalog Number Mfr. No. Material Pore Size Price Quantity    
09-302-100
whatman™
10463808
Regenerated Cellulose-Glass Fiber (RC-GF) 0.45μm Pack of 1000 Each for $3,921.24
09-302-128
whatman™
10463801
Glass Fiber (GF) 1μm Pack of 200 Each for $1,257.87
09-302-129
whatman™
10463800
Glass Fiber (GF) 1μm Pack of 1000 Each for $3,993.28
09-302-131
whatman™
10463815
Glass Fiber (GF) 0.7μm Pack of 1000 Each for $3,921.24
09-302-130
whatman™
10463814
Glass Fiber (GF) 0.7μm Pack of 200 Each for $1,257.87
09-302-132
whatman™
10463803
Nylon (NYL) 0.45μm Pack of 200 Each for $1,348.00
09-302-133
whatman™
10463802
Nylon (NYL) 0.45μm Pack of 1000 Each for $2,847.42
09-302-139
whatman™
10463813
Cellulose Acetate-Glass Fiber (CA-GF) 0.45μm Pack of 200 Each for $1,313.76
09-302-134
whatman™
10463805
Nylon-Glass Fiber (NYL-GF) 0.45μm Pack of 200 Each for $1,334.04
09-302-135
whatman™
10463804
Nylon-Glass Fiber (NYL-GF) 0.45μm Pack of 1000 Each for $3,981.76
09-302-136
whatman™
10463807
Regenerated Cellulose (RC) 0.45μm Pack of 200 Each for $1,031.00
09-302-138
whatman™
10463809
Regenerated Cellulose-Glass Fiber (RC-GF) 0.45μm Pack of 200 Each for $1,313.76
09-302-137
whatman™
10463806
Regenerated Cellulose (RC) 0.45μm Pack of 1000 Each for $3,921.24
Description & Specifications
Specifications
Diameter (Metric) 25mm
Type Syringe Filter
Inlet Style Female luer lock
Outlet Style Male luer
  • Filter housing is made from mechanically stable polypropylene
  • Available with a variety of membrane types; including integrated glass fiber prefilter for difficult samples
  • External geometry of filter housing ensures simple, smooth filter transport from the storage turntable to the filtration site and easy filter changing
  • Suitable for use with Sotax, Caliper, Zymark and Varian tablet testers
  • Connection (In/Out): Luer-lock female/Luer male
Membranes
  • Glass Fiber (GF)
  • Nylon (NL)
  • Regenerated Cellulose (RC)
  • Cellulose Acetate (CA)
Fine filtration of samples in the automatic tablet dissolution test. The Roby 25 Filter Validation Kit can be used for method development.
Note: 200 packs include 8 tubes with 25 pieces each