Please login to your online account to display your discounted pricing
Whatman™ ReZist 30mm PTFE Syringe Filters
Clarifies aggressive, organic solvents. Whatman™ ReZist 30mm PTFE Syringe Filters include integral, permanently hydrophobic PTFE membranes.
$549.35 - $2,639.57
Specifications
Diameter (Metric) 30mm
Material Polytetrafluoroethylene (PTFE)
Housing Material Polypropylene (PP)
Inlet Style Female Luer lock
Outlet Style Male Luer
View More Specs
Products
Catalog Number Mfr. No. Color Code Pore Size Quantity Price Quantity    
09-302-180
whatman™
10 463 503
White 0.2μm 100Pack Pack of 100 Each for $549.35
09-302-182
whatman™
10 463 513
Green 0.45μm 100Pack Pack of 100 Each for $549.35
09-302-186
whatman™
10 463 533
Gray 5μm 100Pack Pack of 100 Each for $549.35
09-302-187
whatman™
10 463 535
Gray 5μm 500Pack Pack of 500 Each for $2,639.57
09-302-184
whatman™
10 463 523
Yellow 1.0μm 100Pack Pack of 100 Each for $549.35
Description & Specifications
Specifications
Diameter (Metric) 30mm
Material Polytetrafluoroethylene (PTFE)
Housing Material Polypropylene (PP)
Inlet Style Female Luer lock
Outlet Style Male Luer
  • Excellent chemical resistance against standard organic HPLC solvents
  • Used as a venting filter for small vessels
  • Hydrophobic PTFE membrane is laminated with polypropylene
  • Permits optimal utilization of small sample volumes