Whatman™ 13mm ReZist™ PTFE Syringe Filters
Designed to be resistant to organic solvents. Whatman™ 13mm ReZist™ PTFE Syringe Filters are ideal for the clarification of aggressive organic solvents and can also be used as a venting filter for small vessels.
Manufacturer:  Whatman™ 10463703
 View more versions of this product
Catalog No. 09-302-178
Specifications
Description
Filter, Syringe; Whatman; ReZist; PTFE; Moisture barrier when venting; Diameter: 13mm.; Pore Size: 0.2μm; Female luer lock inlet and Mini-tip outlet connection; Color code: White; 100/Pk.
  • Hydrophobic PTFE membrane is laminated with polypropylene
  • Mini-Tip outlet is designed for filtration into very small sample bottles
  • Extremely low dead volume <10μL
  • Optimal utilization of small sample volumes
  • Excellent chemical resistance against standard organic HPLC solvents
  • Polypropylene support
  • Used as a venting filter for small vessels
Description & Specifications
Specifications
Syringe Filter
Polytetrafluoroethylene (PTFE)
Polypropylene
female Luer-Lok
100/Pk.
13mm
White
0.2μm
Mini tip