Please login to your online account to display your discounted pricing
Whatman™ 13mm ReZist™ PTFE Syringe Filters
Designed to be resistant to organic solvents. Whatman™ 13mm ReZist™ PTFE Syringe Filters are ideal for the clarification of aggressive organic solvents and can also be used as a venting filter for small vessels.
$445.47 - $462.37
Specifications
Diameter (Metric) 13mm
Type Syringe Filter
Inlet Style female Luer-Lok
Material Polytetrafluoroethylene (PTFE)
Outlet Style Mini tip
View More Specs
Products
Catalog Number Mfr. No. Color Code Pore Size Price Quantity    
09-302-178
whatman™
10463703
White 0.2μm Pack of 100 Each for $445.47
09-302-179
whatman™
10463713
Green 0.45μm Pack of 100 Each for $462.37
Description & Specifications
Specifications
Diameter (Metric) 13mm
Type Syringe Filter
Inlet Style female Luer-Lok
Material Polytetrafluoroethylene (PTFE)
Outlet Style Mini tip
  • Hydrophobic PTFE membrane is laminated with polypropylene
  • Mini-Tip outlet is designed for filtration into very small sample bottles
  • Extremely low dead volume <10μL
  • Optimal utilization of small sample volumes
  • Excellent chemical resistance against standard organic HPLC solvents
  • Polypropylene support
  • Used as a venting filter for small vessels