missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Whatman™ Qualitative Filter Paper: Grade 597½ Pleated Circles

Pleated to save time and achieve optimal fit in conical filter funnels. Whatman™ Qualitative Filter Paper: Grade 597½ Pleated Circles provide extra surface area to increase total loading capacity.

Manufacturer:  Whatman™ WHT10311844*

 View more versions of this product

Catalog No. 09-924-184


 

Specifications

Description

Paper, Filter; Whatman; Qualitative; Folded Prepleated Grades; Thin filter paper, medium-fast with medium to fine particle retention; Used for many routine analytical applications; Grade 597-1/2 circles; Pore size: 4-7um; 125mm; 100/Pk.

Folded filter is used for many routine analytical applications in various industries. Pleated paper enhances and eases filtering process. 
  • Saves time in repetitive or multiple analyses
  • Decreases overall filtration time; normal slow down due to particulate loading is eliminated
  • Increases total loading capacity
  • Maintains flow rate; contact with funnel is reduced, shape of filter is self-supporting
  • Pleating does not significantly affect technical data; same figures may be used with flat circles
  • Grade 597½ is a medium fast paper with medium to fine particle retention
  • Applications include:
    • Food testing; determination of fat content
    • Removal of carbon dioxide and turbidity from beverages
Description & Specifications

Specifications

100Pack
597½
180μm
Medium to fine
Pleated Circle
125mm
Cellulose
4 to 7μm
Medium Fast