Please login to your online account to display your discounted pricing

Whatman™ Qualitative Filter Papers: Grade 2555½ Pleated Circles


Maintains flow rate with reduced contact between filter and funnel. Whatman™Grade 2555½ Pleated Circles are medium fast filter papers used for the filtration of the mash for the determination of the extract in malt and wort and for removing carbon dioxide from beer.

Manufacturer: whatman™  WHT10313951

 View more versions of this product

Catalog No. 09-924-313

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Format Prepleated Circle
Grade 2555½
Quantity 100Pack
Pore Size (Metric) 12μm
Particle Retention Malt extract, wort, CO2
Material Cellulose
Thickness (Metric) 170μ
Flow Rate Medium Fast
For Use With (Application) Filtration of mash for determination of extract in malt, wort, for removing carbondioxide from beer
Diameter (Metric) 240mm

Cellulose filters are used in qualitative analytical techniques to determine and identify materials. Pleated qualitative filters determine and identify materials with improved flow rate and increased loading capacity compared to equivalent quadrant folded filters.

  • Increased filter surface area increases total loading capacity and decreases overall filtration time.
  • Reduced contact between filter and funnel maintains flow rate.
  • Eliminated need to quadrant-fold filter circles to fit conical funnels saves time in repetitive or multiple analyses.
  • Pleated filters have the same technical properties as the respective filter circles.
  • Cellulose filters are used in qualitative analytical techniques to determine and identify materials.
  • Pleated qualitative filters give improved flow rate and increased loading capacity compared to equivalent quadrant folded filters.