Please login to your online account to display your discounted pricing
Whatman™ 13mm Puradisc™ Sterile PVDF Syringe Filters
Designed for high tensile strength, high solvent resistance, and low protein binding. Whatman™ 13mm Puradisc™ Sterile PVDF Syringe Filters are well-suited for biomedical filtration, sterilization filtration, and HPLC sample preparation.
$325.91 - $325.91
Specifications
Material Polyvinylidene difluoride (PVDF)
Housing Material Polypropylene
View More Specs
Products
Catalog Number Mfr. No. Pore Size Sterile Diameter (Metric) Price Quantity    
09-910-18
whatman™
WHT67911302
0.2μm Yes 13mm Pack of 50 Each for $325.91
09-910-19
whatman™
WHT67911304
0.45μm Yes 13mm Pack of 50 Each for $325.91
Description & Specifications
Specifications
Material Polyvinylidene difluoride (PVDF)
Housing Material Polypropylene
  • PVDF filtration media
  • Good resistance to organic solvents
  • Gamma radiation-sterilized
  • Polypropylene housing
  • Available with male Luer or tube-tip outlet (allows direct filtration into microvial inserts)
  • Female Luer lock inlet
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.