missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GE Healthcare Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters

Whatman™ Puradisc™ 25mm Polypropylene Syringe Filters may be used for biological sample and HPLC aqueous sample preparation.

Manufacturer:  GE Healthcare 67862502

 View more versions of this product

Catalog No. 05-713-401



Description

Description

  • Diameter: 25mm
  • Sample volume up to 100mL
  • Low hold-up volumes for maximum sample recovery
  • Optional glass microfiber
  • Autoclave at 121ºC
Applications:
  • HPLC aqueous sample preparation
  • Biological sample preparation
  • Buffer solutions
  • Salt solutions
  • Tissue culture media
  • Irrigation solutions
  • Sterile isolation
Specifications

Specifications

Non-sterile
25 mm
Polypropylene (PP)
Pigment Free Polypropylene (PP)
Up to 100mL
4.2 cm2
Male luer
Puradisc 25 Polypropylene Syringe Filter, 0.2 μm (50 pcs)
22.9 x 28.4 mm
Syringe Filter
25mm
5.2 bar (75 psi)
Whatman Puradisc syringe filters from GE Healthcare’s Life Sciences business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of samples up to 100 mL.
0.2μm
Female luer lock
50
Up to 100 mL
50 Pieces
Documents