Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.45μm BA85 Protran Circles
100% pure nitrocellulose to ensure the highest binding capacity possible
Manufacturer:  Whatman™ WHT10401164
 View more versions of this product
Catalog No. 09-301-105
Specifications Description & Specifications
Specifications
50/Pk.
Nitrocellulose
87mm
0.45μm
Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection
High
Circle