Cytiva Whatman™ PolyVENT™ Vessel Venting Filters
Flows in both directions with hydrophobic PTFE membrane. Whatman™ PolyVENT™ Vessel Venting Filters are integral filter products for sterile venting of vessels and tanks.
$123.87 - $1,968.89
Specifications
Pore Size 0.2 μm
Autoclavable Autoclavable
For Use With (Application) Sterile venting of filling, mixing, holding, autoclave, fermentation, shipping, and dispensing vessels; For isolation of incubators, autoclaves, lyophilizers, EtO sterilizers, and fermenters; For biotech, pharmaceutical and diagnostics, and food and beverage
Material (Membrane) PTFE
View More Specs
Products 5
Catalog Number Mfr. No. Length (Metric) Filtration Area Price Quantity  
Catalog Number Mfr. No. Length (Metric) Filtration Area Price Quantity  
09-745-1D
View Documents
Cytiva
67135036
48 mm 500 cm2
Each for $123.87
09-745-1B
View Documents
Cytiva
67131650
46 mm 16 cm2
Pack of 10 Each for $191.82
09-745-1A
View Documents
Cytiva
67130425
23 mm 4 cm2
Pack of 50 Each for $399.48
09-745-1E
View Documents
Cytiva
67131075
96 mm 1000 cm2
Each for $264.72
09-924-560
View Documents
Cytiva
67131651
46 mm 16 cm2
Pack of 100 Each for $1,968.89
Description
Description
Flows in both directions with hydrophobic PTFE membrane. Whatman™ PolyVENT™ Vessel Venting Filters are integral filter products for sterile venting of vessels and tanks.
Constructed from a single, standardized set of materials.
  • Membrane made of hydrophobic 0.2 µm PTFE membrane.
  • Validated for 50 steam autoclave cycles; compatible with EtO.
  • Testable by Water Break Through (WBT) test or bubble point testing.
  • Pass USP Class VI biosafety tests for plastics.
  • Manufactured in clean room facilities.
  • Bidirectional and autoclavable.
Compliance
  • Biosafe—all materials meet USP Class VI requirements
  • Manufactured in GMP-controlled cleanroom facilities
Specifications
Specifications
0.2 μm
Autoclavable
Sterile venting of filling, mixing, holding, autoclave, fermentation, shipping, and dispensing vessels; For isolation of incubators, autoclaves, lyophilizers, EtO sterilizers, and fermenters; For biotech, pharmaceutical and diagnostics, and food and beverage
PTFE
SDS