Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ Polyethersulfone (PES) Membrane Filters
Whatman polyethersulfone (PES) membrane filters from Cytiva’s Life Sciences business are hydrophilic and stable in alkaline pH, making them suitable for biological samples and aqueous applications.
Manufacturer:  Cytiva (Formerly GE Healthcare Life Sciences) 111164
Catalog No. 09-927-151
Description
Description
  • Low protein binding minimizes nonspecific protein adsorption
  • Hydrophilic membrane suitable for aqueous applications, especially in alkaline pH
  • Smooth surface for easy enumeration of microorganisms
Specifications
Specifications
0.8µm
47mm