Please login to your online account to display your discounted pricing
Whatman™ Polyester Drain Discs
Accessories to membrane filters for support
Manufacturer: whatman™  230300
 View more versions of this product
Catalog No. CS230300
  • / Case of 100 Each
Description & Specifications
Specifications
Material Polyester
Disc Diameter 10mm
Thickness (Metric) 100μm