GE Healthcare Whatman™ Polyester Drain Discs
Provides a flat surface to eliminate filter tearing or rupturing. Whatman™ Polyester Drain Discs are used to support membrane filters. They are available in a variety of diameters for use in a range of devices.
Manufacturer:  GE Healthcare 230300
 View more versions of this product
Catalog No. 05-715-116
Description
Description
For use with membrane hardware where extra support is needed for improved flow rate and throughput. Can be used as a separator between membranes in serial stack filtration applications.
  • Chemically inert polyester
  • 100μm Thickness
  • Binder-free
  • Available in several diameters
Specifications
Specifications
Polyester
Membrane filtration, stack filtration
10mm
100μm
Documents